Learn More
Analytical Sales & Services Lancer C8 Microbore Column. 120Â, 5µm, 5cm x 1.0mm ID
Catalog No. 5000805903
Encompass
Lancer C8 Microbore Column. 120Â, 5um, 5cm x 1.0mm ID
Catalog No. 50-008-05903 Supplier Analytical Sales & Services Supplier No. ADV05L1L000
Lancer C8 Microbore Column. 120Â, 5um, 5cm x 1.0mm ID