Learn More
Analytical Sales & Services Lancer C8 PEEK Guard Cartridge, 5µm, 20mm x 2.1mm ID
Catalog No. 5000805841
Encompass
Lancer C8 PEEK Guard Cartridge, 5µm, 20mm x 2.1mm ID
Catalog No. 50-008-05841 Supplier Analytical Sales & Services Supplier No. ADVH8185
Lancer C8 PEEK Guard Cartridge, 5µm, 20mm x 2.1mm ID