Learn More
Analytical Sales & Services Lancer C8 Prep Column. 120Â, 5µm, 10cm x 20mm ID
Catalog No. 5000806714
Encompass
Lancer C8 Prep Column. 120Â, 5µm, 10cm x 20mm ID
Catalog No. 50-008-06714 Supplier Analytical Sales & Services Supplier No. ADV621020
Lancer C8 Prep Column. 120Â, 5µm, 10cm x 20mm ID