Learn More
Analytical Sales & Services Lancer C8 Prep Column. 120Â, 5µm, 15cm x 20mm ID
Catalog No. 5000805874
Encompass
Lancer C8 Prep Column. 120Â, 5µm, 15cm x 20mm ID
Catalog No. 50-008-05874 Supplier Analytical Sales & Services Supplier No. ADV621520
Lancer C8 Prep Column. 120Â, 5µm, 15cm x 20mm ID