Learn More
Analytical Sales & Services Lancer C8 Prep Column. 120Â, 5µm, 25cm x 20mm ID
Catalog No. 5000806659
Encompass
Lancer C8 Prep Column. 120Â, 5µm, 25cm x 20mm ID
Catalog No. 50-008-06659 Supplier Analytical Sales & Services Supplier No. ADV622520
Lancer C8 Prep Column. 120Â, 5µm, 25cm x 20mm ID