Learn More
Analytical Sales & Services Lancer C8 Semi-Prep Column. 120Â, 5µm, 15cm x 10mm ID
Catalog No. 5000806088
Encompass
Lancer C8 Semi-Prep Column. 120Â, 5µm, 15cm x 10mm ID
Catalog No. 50-008-06088 Supplier Analytical Sales & Services Supplier No. ADV621510
Lancer C8 Semi-Prep Column. 120Â, 5µm, 15cm x 10mm ID