Learn More
Analytical Sales & Services Lancer C8 Semi-Prep Column. 5µm, 25cm x 10mm ID
Catalog No. 5000806081
Encompass
Lancer C8 Semi-Prep Column. 120Â, 5µm, 25cm x 10mm ID
Catalog No. 50-008-06081 Supplier Analytical Sales & Services Supplier No. ADV622510
Lancer C8 Semi-Prep Column. 120Â, 5µm, 25cm x 10mm ID