Learn More
Analytical Sales & Services Lancer Cyano Analytical Column. 120Â, 5µm, 10cm x 2.1mm ID
Catalog No. 5000805964
Encompass
Lancer Cyano Analytical Column. 120Â, 5µm, 10cm x 2.1mm ID
Catalog No. 50-008-05964 Supplier Analytical Sales & Services Supplier No. ADV4021
Lancer Cyano Analytical Column. 120Â, 5µm, 10cm x 2.1mm ID