Learn More
Analytical Sales & Services Lancer Cyano Analytical Column. 120Â, 5µm, 10cm x 4.6mm ID
Catalog No. 5000806149
Encompass
Lancer Cyano Analytical Column. 120Â, 5µm, 10cm x 4.6mm ID
Catalog No. 50-008-06149 Supplier Analytical Sales & Services Supplier No. ADV4011
Lancer Cyano Analytical Column. 120Â, 5µm, 10cm x 4.6mm ID