Learn More
Analytical Sales & Services Lancer Cyano Analytical Column. 120Â, 5µm, 25cm x 4.6mm ID
Catalog No. 5000805597
Encompass
Lancer Cyano Analytical Column. 120Â, 5µm, 25cm x 4.6mm ID
Catalog No. 50-008-05597 Supplier Analytical Sales & Services Supplier No. ADV4013
Lancer Cyano Analytical Column. 120Â, 5µm, 25cm x 4.6mm ID