Learn More
Analytical Sales & Services Lancer Cyano Analytical Column. 120Â, 5µm, 3cm x 3mm ID
Catalog No. 5000805678
Encompass
Lancer Cyano Analytical Column. 120Â, 5µm, 3cm x 3mm ID
Catalog No. 50-008-05678 Supplier Analytical Sales & Services Supplier No. ADV4014
Lancer Cyano Analytical Column. 120Â, 5µm, 3cm x 3mm ID