Learn More
Analytical Sales & Services Lancer Cyano Analytical Column. 120Â, 5µm, 3cm x 4.6mm ID
Catalog No. 5000805965
Encompass
Lancer Cyano Analytical Column. 120Â, 5µm, 3cm x 4.6mm ID
Catalog No. 50-008-05965 Supplier Analytical Sales & Services Supplier No. ADV4009
Lancer Cyano Analytical Column. 120Â, 5µm, 3cm x 4.6mm ID