Learn More
Analytical Sales & Services Lancer Cyano Microbore Column. 120Â, 5µm, 10cm x 0.3mm ID
Catalog No. 5000805904
Encompass
Lancer Cyano Microbore Column. 120Â, 5µm, 10cm x 0.3mm ID
Catalog No. 50-008-05904 Supplier Analytical Sales & Services Supplier No. ADV10C3C000
Lancer Cyano Microbore Column. 120Â, 5µm, 10cm x 0.3mm ID