Learn More
Analytical Sales & Services Lancer Cyano Microbore Column. 120Â, 5µm, 10cm x 0.5mm ID
Catalog No. 5000805865
Encompass
Lancer Cyano Microbore Column. 120Â, 5µm, 10cm x 0.5mm ID
Catalog No. 50-008-05865 Supplier Analytical Sales & Services Supplier No. ADV10C5C000
Lancer Cyano Microbore Column. 120Â, 5µm, 10cm x 0.5mm ID