Learn More
Analytical Sales & Services Lancer Cyano Microbore Column. 120Â, 5µm, 15cm x 1.0mm ID
Catalog No. 5000805953
Encompass
Lancer Cyano Microbore Column. 120Â, 5µm, 15cm x 1.0mm ID
Catalog No. 50-008-05953 Supplier Analytical Sales & Services Supplier No. ADV15C1C000
Lancer Cyano Microbore Column. 120Â, 5µm, 15cm x 1.0mm ID