Learn More
Analytical Sales & Services Lancer Cyano Microbore Column. 120Â, 5µm, 5cm x 0.3mm ID
Catalog No. 5000805924
Encompass
Lancer Cyano Microbore Column. 120Â, 5µm, 5cm x 0.3mm ID
Catalog No. 50-008-05924 Supplier Analytical Sales & Services Supplier No. ADV05C3C000
Lancer Cyano Microbore Column. 120Â, 5µm, 5cm x 0.3mm ID