Learn More
Analytical Sales & Services Lancer Cyano Microbore Column. 120Â, 5µm, 5cm x 0.5mm ID
Catalog No. 5000805863
Encompass
Lancer Cyano Microbore Column. 120Â, 5µm, 5cm x 0.5mm ID
Catalog No. 50-008-05863 Supplier Analytical Sales & Services Supplier No. ADV05C5C000
Lancer Cyano Microbore Column. 120Â, 5µm, 5cm x 0.5mm ID