Learn More
Analytical Sales & Services Lancer Cyano Microbore Column. 120Â, 5µm, 5cm x 1.0mm ID
Catalog No. 5000805659
Encompass
Lancer Cyano Microbore Column. 120Â, 5µm, 5cm x 1.0mm ID
Catalog No. 50-008-05659 Supplier Analytical Sales & Services Supplier No. ADV05C1C000
Lancer Cyano Microbore Column. 120Â, 5µm, 5cm x 1.0mm ID