Learn More
Analytical Sales & Services Lancer Phenyl Analytical Column. 120Â, 4.5µm, 10cm x 4.6mm ID
Catalog No. 5000805999
Encompass
Lancer Phenyl Analytical Column. 120Â, 4.5µm, 10cm x 4.6mm ID
Catalog No. 50-008-05999 Supplier Analytical Sales & Services Supplier No. ADV5906
Lancer Phenyl Analytical Column. 120Â, 4.5µm, 10cm x 4.6mm ID