Learn More
Analytical Sales & Services Lancer Phenyl Analytical Column. 120Â, 4.5µm, 15cm x 3.0mm ID
Catalog No. 5000805767
Encompass
Lancer Phenyl Analytical Column. 120Â, 4.5µm, 15cm x 3.0mm ID
Catalog No. 50-008-05767 Supplier Analytical Sales & Services Supplier No. ADV5907
Lancer Phenyl Analytical Column. 120Â, 4.5µm, 15cm x 3.0mm ID