Learn More
Analytical Sales & Services Lancer Phenyl Analytical Column. 120Â, 4.5µm, 15cm x 4.6mm ID
Catalog No. 5000805478
Encompass
Lancer Phenyl Analytical Column. 120Â, 4.5µm, 15cm x 4.6mm ID
Catalog No. 50-008-05478 Supplier Analytical Sales & Services Supplier No. ADV5912
Lancer Phenyl Analytical Column. 120Â, 4.5µm, 15cm x 4.6mm ID