Learn More
Analytical Sales & Services Lancer Phenyl Analytical Column. 120Â, 4.5µm, 5cm x 3.0mm ID
Catalog No. 5000806001
Encompass
Lancer Phenyl Analytical Column. 120Â, 4.5µm, 5cm x 3.0mm ID
Catalog No. 50-008-06001 Supplier Analytical Sales & Services Supplier No. ADV5910
Lancer Phenyl Analytical Column. 120Â, 4.5µm, 5cm x 3.0mm ID