Learn More
Analytical Sales & Services Lancer Phenyl Microbore Column. 120Â, 4.5µm, 10cm x 1.0mm ID
Catalog No. 5000804996
Encompass
Lancer Phenyl Microbore Column. 120Â, 4.5µm, 10cm x 1.0mm ID
Catalog No. 50-008-04996 Supplier Analytical Sales & Services Supplier No. ADV10P1P000
Add to Cart
Add to Cart
Lancer Phenyl Microbore Column. 120Â, 4.5µm, 10cm x 1.0mm ID