Learn More
Analytical Sales & Services Lancer Phenyl Microbore Column. 120Â, 4.5µm, 15cm x 0.5mm ID
Catalog No. 5000805677
Encompass
Lancer Phenyl Microbore Column. 120Â, 4.5µm, 15cm x 0.5mm ID
Catalog No. 50-008-05677 Supplier Analytical Sales & Services Supplier No. ADV15P5P000
Add to Cart
Add to Cart
Lancer Phenyl Microbore Column. 120Â, 4.5µm, 15cm x 0.5mm ID