Learn More
Analytical Sales & Services Lancer Phenyl Microbore Column. 120Â, 4.5µm, 25cm x 1.0mm ID
Catalog No. 5000806706
Encompass
Lancer Phenyl Microbore Column. 120Â, 4.5µm, 25cm x 1.0mm ID
Catalog No. 50-008-06706 Supplier Analytical Sales & Services Supplier No. ADV25P1P000
Lancer Phenyl Microbore Column. 120Â, 4.5µm, 25cm x 1.0mm ID