Learn More
Analytical Sales & Services Lancer Phenyl Microbore Column. 120Â, 4.5µm, 3cm x 0.3mm ID
Catalog No. 5000805986
Encompass
Lancer Phenyl Microbore Column. 120Â, 4.5µm, 3cm x 0.3mm ID
Catalog No. 50-008-05986 Supplier Analytical Sales & Services Supplier No. ADV03P3P000
Lancer Phenyl Microbore Column. 120Â, 4.5µm, 3cm x 0.3mm ID