Learn More
Analytical Sales & Services Lancer Phenyl Microbore Column. 120Â, 4.5µm, 5cm x 1.0mm ID
Catalog No. 5000805951
Encompass
Lancer Phenyl Microbore Column. 120Â, 4.5µm, 5cm x 1.0mm ID
Catalog No. 50-008-05951 Supplier Analytical Sales & Services Supplier No. ADV05P1P000
Lancer Phenyl Microbore Column. 120Â, 4.5µm, 5cm x 1.0mm ID