Learn More
Analytical Sales & Services Lancer SS Guard Cartridge, C18, 120Â, 5µm, 20mm x 3.2mm ID
Catalog No. 5000806903
Encompass
Lancer Stainless Steel Guard Cartridge, C18, 120Â, 5µm, 20mm x 3.2mm ID
Catalog No. 50-008-06903 Supplier Analytical Sales & Services Supplier No. ADVH218
Lancer Stainless Steel Guard Cartridge, C18, 120Â, 5µm, 20mm x 3.2mm ID