Learn More
Medchemexpress LLC HY-N2159 5mg Medchemexpress, Lancerin CAS:81991-99-3 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYN21595MG
Encompass_Preferred
Medchemexpress, HY-N2159 5mg Lancerin CAS:81991-99-3 Lancerin, isolated from the root bark of Cudraniu cochinchinensis, possesses anti-lipid peroxidation. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2870