Learn More
Med Vet International Safety Lancet McKesson 26 Gauge Retractable Pressure Activated Finger, 16-PASL26GX, 100/Box, 20 Boxes
Catalog No. 503043150
Encompass_Preferred
Blue in color, defines a 26 Gauge needle Sterile tri-bevel needle for added protection Depth setting of 2.0 mm Lancets are ready to use, no loading required After use needle automatically retracts into housing preventing needlestick injuries
Catalog No. 50-304-3150 Supplier Med Vet International Supplier No. 1217982CS
Blue in color, defines a 26 Gauge needle Sterile tri-bevel needle for added protection Depth setting of 2.0 mm Lancets are ready to use, no loading required After use needle automatically retracts into housing preventing needlestick injuries