Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sarstedt Inc Safety Lancet, 1.8mm x 0.8mm, 200/Pack

Catalog No. NC0165197 Shop All Sarstedt Inc. Products
Encompass_Preferred

Safety Lancet 1.8mm x 0.8mm Super

Catalog No. NC0165197 Supplier Sarstedt Inc Supplier No. 85.1018
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Safety Lancet 1.8mm x 0.8mm Super

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.