Learn More
Med Vet International Safety Lancet McKesson 21 Gauge Retractable Pressure Activated Finger, 16-PASL21GX, 100/Box
Catalog No. 503044777
Encompass_Preferred
Green in color, defines a 21 Gauge needle Sterile tri-bevel needle for added protection Depth setting of 2.0 mm Lancets are ready to use, no loading required After use needle automatically retracts into housing preventing needlestick injuries
Catalog No. 50-304-4777 Supplier Med Vet International Supplier No. 1217981BX
Green in color, defines a 21 Gauge needle Sterile tri-bevel needle for added protection Depth setting of 2.0 mm Lancets are ready to use, no loading required After use needle automatically retracts into housing preventing needlestick injuries