Learn More
Med Vet International Safety Lancet McKesson 21 Gauge Retractable Pressure Activated Finger, 16-PASL21GX, 100/Box, 20 Boxes
Catalog No. 503044776
Encompass_Preferred
Green in color, defines a 21 Gauge needle Sterile tri-bevel needle for added protection Depth setting of 2.0 mm Lancets are ready to use, no loading required After use needle automatically retracts into housing preventing needlestick injuries
Catalog No. 50-304-4776 Supplier Med Vet International Supplier No. 1217981CS
Green in color, defines a 21 Gauge needle Sterile tri-bevel needle for added protection Depth setting of 2.0 mm Lancets are ready to use, no loading required After use needle automatically retracts into housing preventing needlestick injuries