Learn More
Bound Tree Medical LANCET FOR USE WITH GLUCOMETER
Supplier:  Bound Tree Medical 675AT1062
Encompass_Preferred
Unistik� 3 Lancet needle point is hidden before use and automatically retracts after use to avoid accidental needlestick injuries and cross infection
Catalog No. NC0653145