Learn More
Med Vet International Safety Lancet Surgilance 21 Gauge Retractable Push Button Activation Finger, SLN240, 100/Box
Catalog No. 503044554
Encompass_Preferred
Safe, Easy & Effective Taking patient samples has never been easier Low, medium and high blood flow lancets make SurgiLance safety lancets the ideal choice for all your testing requirements Needle or blade is safely concealed before and after use Once the lancet has been used, it is rendered inoperable, providing added safety for both clinician and patient No arming required Lancets are color-coded for easy recognition High-speed delivery minimizes clinician error and patient pain
Catalog No. 50-304-4554 Supplier Med Vet International Supplier No. 511848BX
Safe, Easy & Effective Taking patient samples has never been easier Low, medium and high blood flow lancets make SurgiLance safety lancets the ideal choice for all your testing requirements Needle or blade is safely concealed before and after use Once the lancet has been used, it is rendered inoperable, providing added safety for both clinician and patient No arming required Lancets are color-coded for easy recognition High-speed delivery minimizes clinician error and patient pain