Learn More
Med Vet International McKesson Twist Lancet, 30G, 16-030-100X, 100/Box
Catalog No. 503043193
Encompass_Preferred
Tri-bevel tip Consistent depth penetration Universal design fits most lancing devices Designed to facilitate virtually painless sampling Not made with natural rubber latex
Catalog No. 50-304-3193 Supplier Med Vet International Supplier No. 1217979BX
Tri-bevel tip Consistent depth penetration Universal design fits most lancing devices Designed to facilitate virtually painless sampling Not made with natural rubber latex