Learn More
Med Vet International Safety Lancet Assure 30 Gauge Protective Safety Cap Push Button Activation Finger, 990130, 100/Box
Catalog No. 503045417
Encompass_Preferred
The hidden-needle construction and automatic after-use lockout helps prevent needlestick accidents Firing Button: Easier to engage while reducing the likelihood of a misfire Shallow Pen Depth: Reduces the amount of discomfort Ergonomic Shape: Provides secure device handling
Catalog No. 50-304-5417 Supplier Med Vet International Supplier No. 962941BX
The hidden-needle construction and automatic after-use lockout helps prevent needlestick accidents Firing Button: Easier to engage while reducing the likelihood of a misfire Shallow Pen Depth: Reduces the amount of discomfort Ergonomic Shape: Provides secure device handling