Learn More
Med Vet International Dynarex Pressure ActIVated Safety Lancets - Sterile, 28g, 1.8 mm, 1000/Case
Catalog No. 502829572
Encompass_Preferred
The Dynarex Safety Lancets are high-quality, medical-grade lancets designed for use in capillary blood sampling, diagnostic testing, and individual blood sugar monitoring. The lancets are pre-loaded, eliminating the need for additional lancing devices. The sterile tri-beveled needle features a retractable protective sheath to reduce the risk of needlestick injuries. Available in a variety of gauge sizes.
Catalog No. 50-282-9572 Supplier Med Vet International Supplier No. 7115
The Dynarex Safety Lancets are high-quality, medical-grade lancets designed for use in capillary blood sampling, diagnostic testing, and individual blood sugar monitoring. The lancets are pre-loaded, eliminating the need for additional lancing devices. The sterile tri-beveled needle features a retractable protective sheath to reduce the risk of needlestick injuries. Available in a variety of gauge sizes.