Learn More
Med Vet International EasyTouch Pressure-Activated Safety Lancets, 23G x 2.2mm, 100/Box
Supplier:  Med Vet International 823221
Encompass_Preferred
Each EasyTouch Pressure Activated Safety Lancets are color-coded by gauge for easy and accurate identification. Confusion is reduced significantly by allowing customers to quickly identify their correct gauge and needle length. Features & Benefits Needle is shielded both prior and after use Easy-to-use button design Triple-Bevel cut for a sharper point No lancing device required Sterilized by Gamma Radiation Color Coded by gauge Bilingual Packaging
Catalog No. 50-255-9653