Learn More
Med Vet International EasyTouch Twist Lancets, 33G, 100/Box, 5 Boxes
Supplier:  Med Vet International 833101X5
Encompass_Preferred
EasyTouch Twist Lancets are designed to fit the most commonly used lancing devices and have a variety of gauges to fit your level of comfort. Sterile, Single use only Fits Most Commonly used Lancing Devices Latex Free Triple-Bevel Cut Bilingual Packaging Color Coded by Gauge (Box & Lancet)
Catalog No. 50-209-2431