Learn More
VivaGuard™ Lancing Device
Catalog No. 23111433
Change view
Click to view available options
For Use With (Equipment):
Germaine™ Lancet
Catalog No. For Use With (Equipment)
23-111-433 Germaine™ Lancet
1 options
This item is not returnable. View return policy
Catalog No. 23-111-433 Supplier VivaGuard™ Supplier No. VGD01381
Add to Cart
Add to Cart
This item is not returnable. View return policy
This lancing device has five depth settings for maximum comfort, a secure grip, smooth gliding mechanism, and safe lancet ejection.
  • VivaGuard™ Lancing Device Lancing Device with five depth settings offers maximum comfort through selecting a proper penetration depth
  • Easy to carry with a secure-grip body design for safety
  • Smooth gliding mechanism offers reliability and simplicity
  • Unique design and easy-to-use
  • Lancet ejector easily disposes of the used lancet, hands-free, to prevent accidents and for better safety
  • Convenient storage with 5-year shelf life
Specifications
Product Type Lancet
Disposable Reusable
For Use With (Application) Fingerstick Sample Tests, Capillary Testing
Sterility Non-sterile
Certifications/Compliance FDA Cleared for OTC and Professional Use
For Use With (Equipment) Germaine™ Lancet
Includes Lancing Device, Instructions