Learn More
Aviva Systems Biology Corp LANCL3 Antibody - C-terminal region (ARP55888_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP55888_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against LANCL3. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the C terminal region of human LANCL3. Alternate names: FLJ42925ApplicationWB. Species reactivity: Cow, Dog, Goat, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-286-092