Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Supertek™ Landform 3D Models
GREENER_CHOICESCI_ED

Catalog No. S81674 Shop All Supertek Scientific Products
Change view
Click to view available options
Dimensions (L x W):
35 x 35 in.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (L x W)
S81674 35 x 35 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S81674 Supplier Supertek™ Supplier No. B19048
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Investigate geographical and geological features

  • Eight 3D landform models showing the mountains, lakes, rivers, deserts, glaciers, coastlines, canyons and cultural point
  • Includes lesson plan
  • Finished size 35 x 35 in.

Specifications

Grade 5+
Product Type Model
Includes Eight 3D landform models and lesson plan
Dimensions (L x W) 35 x 35 in.
For Use With (Application) Teach Landforms
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.