Learn More
Medchemexpress LLC Landiolol hydrochloride | 144481-98-1 | 546.05 | C25H40ClN3O8 | 100 MG
SDP
Catalog No. 5000214833
Encompass_Preferred
Landiolol hydrochloride is an ultra-short-acting, highly selective β1-adrenergic receptor antagonist provided as a research reagent and analytical standard. It is used to study β1 receptor-mediated pharmacology in vitro and in vivo and is supplied with product documentation for safe handling and experimental use.
  • Ultra-short-acting selective β1-adrenergic receptor antagonist.
  • Commonly used as an analytical standard and pharmacological tool.
  • Supplied as a powder suitable for in vitro and in vivo research.
  • Available in multiple pack sizes, including 100 MG.
  • Supplied with a datasheet and safety data sheet (SDS) for handling information.
  • CAS number 144481-98-1 and molecular weight 546.05 g/mol for identification.
Catalog No. 50-002-14833 Supplier Medchemexpress LLC Supplier No. HY100607A100MG
Landiolol hydrochloride is an ultra-short-acting, highly selective β1-adrenergic receptor antagonist provided as a research reagent and analytical standard. It is used to study β1 receptor-mediated pharmacology in vitro and in vivo and is supplied with product documentation for safe handling and experimental use.
  • Ultra-short-acting selective β1-adrenergic receptor antagonist.
  • Commonly used as an analytical standard and pharmacological tool.
  • Supplied as a powder suitable for in vitro and in vivo research.
  • Available in multiple pack sizes, including 100 MG.
  • Supplied with a datasheet and safety data sheet (SDS) for handling information.
  • CAS number 144481-98-1 and molecular weight 546.05 g/mol for identification.