Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Landiolol hydrochloride | 144481-98-1 | MFCD01937430 | >98.0% | 546.05 | C25H40ClN3O8 | 50 MG
SDP

Catalog No. 5000214638
Encompass_Preferred

Landiolol hydrochloride is an analytical standard of an ultra-short-acting, highly β1-selective beta-adrenergic receptor antagonist supplied for research and analytical use, suitable for quantification, calibration, and method development in biochemical and pharmacological studies.

  • Analytical standard grade for research and method development.
  • High purity, >98% (HPLC) as reported by suppliers.
  • Molecular formula C25H40ClN3O8 and molecular weight 546.05 Da.
  • CAS number 144481-98-1 for unambiguous identification.
  • Supplied in small quantities suitable for analytical use, e.g. 50 MG.
  • Intended for calibration, quantification, and analytical characterization in biological and formulation samples.
  • Store under conditions specified in the certificate of analysis.

Catalog No. 50-002-14638 Supplier Medchemexpress LLC Supplier No. HY100607A50MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Landiolol hydrochloride is an analytical standard of an ultra-short-acting, highly β1-selective beta-adrenergic receptor antagonist supplied for research and analytical use, suitable for quantification, calibration, and method development in biochemical and pharmacological studies.

  • Analytical standard grade for research and method development.
  • High purity, >98% (HPLC) as reported by suppliers.
  • Molecular formula C25H40ClN3O8 and molecular weight 546.05 Da.
  • CAS number 144481-98-1 for unambiguous identification.
  • Supplied in small quantities suitable for analytical use, e.g. 50 MG.
  • Intended for calibration, quantification, and analytical characterization in biological and formulation samples.
  • Store under conditions specified in the certificate of analysis.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.