Learn More
Medchemexpress LLC Landiolol hydrochloride | 144481-98-1 | MFCD01937430 | 99.8% | 546.05 g·mol⁻¹ | C25H40ClN3O8 | 25 MG
SDP
Catalog No. 5000214682
Encompass_Preferred
Landiolol hydrochloride is the hydrochloride salt of landiolol, an ultra-short-acting, highly selective β1-adrenergic receptor antagonist used in research to reduce heart rate and investigate cardioprotective and sepsis-related effects. Supplied as a high-purity solid analytical standard, it is intended for in vitro and preclinical studies where transient β1 blockade is required.
  • Highly selective β1-adrenergic receptor antagonist.
  • Ultra-short acting profile for transient β1 blockade.
  • High purity suitable for analytical and preclinical use.
  • Supplied as a solid powder for easy weighing and formulation.
  • Defined storage recommendations to maintain stability.
Catalog No. 50-002-14682 Supplier Medchemexpress LLC Supplier No. HY100607A25MG
Landiolol hydrochloride is the hydrochloride salt of landiolol, an ultra-short-acting, highly selective β1-adrenergic receptor antagonist used in research to reduce heart rate and investigate cardioprotective and sepsis-related effects. Supplied as a high-purity solid analytical standard, it is intended for in vitro and preclinical studies where transient β1 blockade is required.
  • Highly selective β1-adrenergic receptor antagonist.
  • Ultra-short acting profile for transient β1 blockade.
  • High purity suitable for analytical and preclinical use.
  • Supplied as a solid powder for easy weighing and formulation.
  • Defined storage recommendations to maintain stability.