Learn More
TARGETMOL CHEMICALS INC LANDIPIRDINE 5MG
SDP
Catalog No. 502580578
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg and bulk. Please contact Fisher for quotes. Landipirdine (RO-5025181) is a selective 5-HT6R antagonist for the study of central nervous system disorders such as dementia. purity: 99%
Catalog No. 50-258-0578 Supplier TARGETMOL CHEMICALS INC Supplier No. T277955MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg and bulk. Please contact Fisher for quotes. Landipirdine (RO-5025181) is a selective 5-HT6R antagonist for the study of central nervous system disorders such as dementia. purity: 99%