Learn More
Electron Microscopy Sciences Langenbech Metacarpal Saw, 229 mm
SDP
Catalog No. 501927036
Encompass_Preferred
Three styles of manual bone saws are available. The broad bladed Satterlee Saw (A), the thinner rounded tip Charriere Saw (B), and the solid handle Langenbech Metacarpal Saw (C). Each saw has serrated teeth. Solid handle.
OAL: 229 mm
Catalog No. 50-192-7036 Supplier Electron Microscopy Sciences Supplier No. 7056633
Three styles of manual bone saws are available. The broad bladed Satterlee Saw (A), the thinner rounded tip Charriere Saw (B), and the solid handle Langenbech Metacarpal Saw (C). Each saw has serrated teeth. Solid handle.
OAL: 229 mm