Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Stoelting™ Langenbeck Saw
SDP

Catalog No. 10001818 Shop All Stoelting Co Products
Change view
Click to view available options
Size:
Blade: 4.5 in.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
10-001-818 Blade: 4.5 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 10-001-818 Supplier Stoelting™ Supplier No. 5217003
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Bone cutting tools allow for sharp cuts through soft bone and cartilage.

Specifications

For Use With (Application) Neuroscience
Length (English) 9 in.
Size Blade: 4.5 in.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.