Learn More
TARGETMOL CHEMICALS INC Lanicemine dihydrochloride 2
SDP
Catalog No. 502879833
Encompass_Preferred
Also available in 1 mL, 2 mg, 5 mg, 10 mg, 50 mg, 100 mg, 500 mg and bulk. Please contact Fisher for quotes. Lanicemine dihydrochloride (AZD-6765 dihydrochloride) is a low-capture NMDA channel blocker with antidepressant activity and can be used to study neurological diseases. Purity 99.43%
Catalog No. 50-287-9833 Supplier TARGETMOL CHEMICALS INC Supplier No. T6047125MG
Also available in 1 mL, 2 mg, 5 mg, 10 mg, 50 mg, 100 mg, 500 mg and bulk. Please contact Fisher for quotes. Lanicemine dihydrochloride (AZD-6765 dihydrochloride) is a low-capture NMDA channel blocker with antidepressant activity and can be used to study neurological diseases. Purity 99.43%